missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2-Methoxynaphthalene(Nerolin), Mikromol™
Click to view available options
Quantity:
25 mg
100 mg
4 x 25 mg
Unit Size:
100mg
25mg
Description
2-Methoxynaphthalene(Nerolin)
Specifications
Specifications
| Chemical Name or Material | 2-Methoxynaphthalene(Nerolin) |
| CAS | 93-04-9 |
| Analyte or Component Names | 2-Methoxynaphthalene(Nerolin) |
| Shipping Condition | Room Temperature |
| Molecular Formula | C11 H10 O |
| Certifications/Compliance | ISO 17025 |
| Synonym | 2-Methoxynaphthalene(Nerolin), Naproxen Imp. M (EP), Naproxen Impurity M |
| InChI Formula | InChI=1S/C11H10O/c1-12-11-7-6-9-4-2-3-5-10(9)8-11/h2-8H,1H3 |
| SMILES | COc1ccc2ccccc2c1 |
| IUPAC Name | 2-methoxynaphthalene |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction