missing translation for 'onlineSavingsMsg'
Learn More

2-Methoxynaphthalene, 98%

Product Code. 10377530 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
5 g
500 g
Packaging:
Glass bottle
Plastic bottle
Unit Size:
500g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10377530 500 g Plastic bottle 500g
10791241 5 g Glass bottle 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 missing translation for 'options'
This item is not returnable. View return policy
Product Code. 10377530 missing translation for 'mfr' Thermo Scientific Acros missing translation for 'supplierNo' 164755000
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 93-04-9 | C11H10O | 158.2 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 93-04-9
Molecular Formula C11H10O
Molecular Weight (g/mol) 158.2
MDL Number MFCD00004061
InChI Key LUZDYPLAQQGJEA-UHFFFAOYSA-N
Synonym naphthalene, 2-methoxy, yara yara, nerolin, methyl 2-naphthyl ether, yura yara, 2-naphthyl methyl ether, yara-yara, 2-naphthol methyl ether, nerolin old, beta-naphthyl methyl ether
PubChem CID 7119
IUPAC Name 2-methoxynaphthalene
SMILES COC1=CC2=CC=CC=C2C=C1

Specifications

CAS Min % 97.5
CAS Max % 100.0
Melting Point 73°C to 75°C
Boiling Point 274°C
Infrared Spectrum Authentic
Assay Percent Range 98%
Packaging Plastic bottle
Linear Formula C10H7OCH3
Quantity 500 g
Beilstein 06, 640
Merck Index 15, 6070
Solubility Information Solubility in water: insoluble in water. Other solubilities: soluble in alcohol,ether,carbon disulfide and,benzene
Formula Weight 158.2
Percent Purity 98%
Physical Form Crystalline Solid
Chemical Name or Material 2-Methoxynaphthalene, 98%
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.