missing translation for 'onlineSavingsMsg'
Learn More

2-(Methoxymethyl)phenylboronic acid, 98%, Thermo Scientific™

Product Code. 15244368 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
Product Code. Quantity unitSize
15244368 5 g 5g
15234368 1 g 1g
2 options
This item is not returnable. View return policy

Product Code. 15244368

Brand: Thermo Scientific Acros 454550050

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Chemical Identifiers

CAS 126617-98-9
Molecular Formula C8H11BO3
Molecular Weight (g/mol) 165.99
InChI Key JHPPAFXQJZJGEP-UHFFFAOYSA-N
Synonym 2-methoxymethylphenylboronic acid, 2-methoxymethylbenzeneboronic acid, 2-methoxymethyl phenyl boronic acid, 2-methoxymethyl phenylboronic acid, 2-methoxymethyl benzeneboronic acid, 2-methoxymethylphebylboronic acid, boronic acid, 2-methoxymethyl phenyl, pubchem7997
PubChem CID 3759408
IUPAC Name [2-(methoxymethyl)phenyl]boronic acid
SMILES B(C1=CC=CC=C1COC)(O)O

Specifications

Melting Point 42.0°C to 44.0°C
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (HPLC)
Quantity 5 g
Formula Weight 165.99
Percent Purity 98%
Chemical Name or Material 2-(Methoxymethyl)phenylboronic acid
compliance-icons
Product Identifier
  • 2-(Methoxymethyl)phenylboronic acid, 98%
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
Hazard Statement
  • H312-Harmful in contact with skin.
  • H332-Harmful if inhaled.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.