missing translation for 'onlineSavingsMsg'
Learn More

2-Methoxyfuran, 97%

Product Code. 11427447
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
Unit Size:
1g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11427447 5 g 5g
11417447 1 g 1g
11437447 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11427447 Supplier Thermo Scientific Alfa Aesar Supplier No. A15593.06
 Restricted Item
In Stock
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 25414-22-6 | C5H6O2 | 98.101 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 25414-22-6
Molecular Formula C5H6O2
Molecular Weight (g/mol) 98.101
MDL Number MFCD00003227
InChI Key OXCGHDNCMSOEBZ-UHFFFAOYSA-N
PubChem CID 117476
IUPAC Name 2-methoxyfuran
SMILES COC1=CC=CO1

Specifications

Density 1.065
Boiling Point 110°C to 111°C
Flash Point 10°C (50°F)
Refractive Index 1.447
Quantity 5 g
UN Number UN3271
Beilstein 106270
Synonym furan, 2-methoxy, 2-methoxy furan, unii-25y8i1p35t, 2-furyl methyl ether, furan,2-methoxy, pubchem19998, acmc-209gjs, 2-methoxyfuran, 2-furyl methyl ether #
Formula Weight 98.1
Percent Purity 97%
Chemical Name or Material 2-Methoxyfuran
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.