missing translation for 'onlineSavingsMsg'
Learn More

2-Methoxyfuran, 97%

Product Code. 11417447
Click to view available options
Quantity:
1 g
5 g
25 g
Unit Size:
1g
25g
5g
This item is not returnable. View return policy

Product Code. 11417447

Brand: Thermo Scientific Alfa Aesar A15593.03

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 25414-22-6 | C5H6O2 | 98.101 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 25414-22-6
Molecular Formula C5H6O2
Molecular Weight (g/mol) 98.101
MDL Number MFCD00003227
InChI Key OXCGHDNCMSOEBZ-UHFFFAOYSA-N
PubChem CID 117476
IUPAC Name 2-methoxyfuran
SMILES COC1=CC=CO1

Specifications

Density 1.065
Boiling Point 110°C to 111°C
Flash Point 10°C (50°F)
Refractive Index 1.447
Quantity 1 g
UN Number UN3271
Beilstein 106270
Synonym furan, 2-methoxy, 2-methoxy furan, unii-25y8i1p35t, 2-furyl methyl ether, furan,2-methoxy, pubchem19998, acmc-209gjs, 2-methoxyfuran, 2-furyl methyl ether #
Formula Weight 98.1
Percent Purity 97%
Chemical Name or Material 2-Methoxyfuran
Show More Show Less

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.