missing translation for 'onlineSavingsMsg'
Learn More

2-Methoxyethyl phenyl sulfide, 98%, Thermo Scientific™

Product Code. 11328445
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11328445 25 g 25g
11318445 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11328445 Supplier Thermo Scientific Alfa Aesar Supplier No. L10464.14
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 77298-24-9
Molecular Formula C9H12OS
Molecular Weight (g/mol) 168.25
MDL Number MFCD00041329
InChI Key ONVZBVAUSAELNQ-UHFFFAOYSA-N
Synonym 2-methoxyethyl phenyl sulfide, 2-methoxyethyl phenyl sulfane, 2-methoxyethyl sulfanyl benzene, 2-methoxyethyl phenylsulfide, 1-methoxy-2-phenylthioethane, 2-phenylthio ethyl methyl ether, benzene, 2-methoxyethyl thio
PubChem CID 4056174
IUPAC Name 2-methoxyethylsulfanylbenzene
SMILES COCCSC1=CC=CC=C1

Specifications

Boiling Point 125°C to 126°C (18 mmHg)
Linear Formula C6H5S(CH2)2OCH3
Refractive Index 1.557
Quantity 25 g
Beilstein 5500488
Formula Weight 168.25
Percent Purity 98%
Chemical Name or Material 2-Methoxyethyl phenyl sulfide
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.