missing translation for 'onlineSavingsMsg'
Learn More

2-Methoxyethanol, ReagentPlus™, ≥99.0%, Honeywell™

Product Code. p-7120067 Shop All Honeywell Research Chemicals Products
Click to view available options
Quantity:
1 L
2.5 L
20 L
Packaging:
Glass Bottle
Stainless Steel Can
Unit Size:
1 L
2.5 L
20 L
This item is not returnable. View return policy

Product Code. 15601680

Brand: Honeywell Research Chemicals 1854691L

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Reagent Grade, contains 50 ppm BHT as stabilizer,≥99.0%

  • ≥95% for general laboratory use.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 109-86-4
Molecular Formula C3H8O2
Molecular Weight (g/mol) 76.095
MDL Number MFCD00002867
InChI Key XNWFRZJHXBZDAG-UHFFFAOYSA-N
Synonym methyl cellosolve, ethanol, 2-methoxy, ethylene glycol monomethyl ether, methyl oxitol, 2-methoxy-1-ethanol, methoxyethanol, 3-oxa-1-butanol, egme, monomethyl glycol, dowanol em
PubChem CID 8019
ChEBI CHEBI:46790
IUPAC Name 2-methoxyethanol
SMILES COCCO

Specifications

Melting Point -85°C
Color Colorless
Density 0.965g/mL at 25°C
pH 4 to 7
Boiling Point 124°C
Explosion Limit 0.245
Flash Point 40°C (104°F)
Ignition Point 548°F
Packaging Glass Bottle
Linear Formula CH3OCH2CH2OH
Refractive Index n20/D 1.402(lit.)
Quantity 1 L
UN Number UN1188
Beilstein 1731074
Formula Weight 76.09g/mol
Vapor Pressure 6.17mmHg (20°C)
Percent Purity ≥99.0%
Grade ReagentPlus
Physical Form Liquid
Chemical Name or Material 2-Methoxyethanol
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.