Learn More
2-Methoxyethanol, Extra Pure, SLR, Fisher Chemical™
Brand: Fisher Chemical M/4100/PB17
Code : 21
Additional Details : CAS Number : 109-86-4 Weight : 2.41250kg Transport : UN number : 1188 Chem class : 3 Pack group : III
Packaging | Plastic Coated Glass bottle |
---|---|
Quantity | 2.5L |
Chemical Identifiers
109-86-4 | |
76.095 | |
XNWFRZJHXBZDAG-UHFFFAOYSA-N | |
8019 | |
2-methoxyethanol |
C3H8O2 | |
2867 | |
methyl cellosolve, ethanol, 2-methoxy, ethylene glycol monomethyl ether, methyl oxitol, 2-methoxy-1-ethanol, methoxyethanol, 3-oxa-1-butanol, egme, monomethyl glycol, dowanol em | |
CHEBI:46790 | |
COCCO |
Specifications
Plastic Coated Glass bottle | |
124°C | |
-85°C | |
109-86-4 | |
2867 | |
XNWFRZJHXBZDAG-UHFFFAOYSA-N | |
2-methoxyethanol | |
8019 | |
76.09g/mol |
9.5mmHg at 25°C | |
Colorless | |
2.5L | |
C3H8O2 | |
methyl cellosolve, ethanol, 2-methoxy, ethylene glycol monomethyl ether, methyl oxitol, 2-methoxy-1-ethanol, methoxyethanol, 3-oxa-1-butanol, egme, monomethyl glycol, dowanol em | |
COCCO | |
76.095 | |
CHEBI:46790 | |
Liquid |
Safety and Handling
- 2-Methoxyethanol
Signal Word
- Danger
Hazard Category
- Acute toxicity Category 4
- Flammable liquid Category 3
- Reproductive toxicity Category 1B
- Specific target organ toxicity after repeated exposure Category 2
- Specific target organ toxicity after single exposure Category 1
Hazard Statement
- H370-Causes damage to organs.
- H360FD-May damage fertility. May damage the unborn child.
- H373-May cause damage to organs.
- H302-Harmful if swallowed.
- H312-Harmful in contact with skin.
- H332-Harmful if inhaled.
- H226-Flammable liquid and vapour.
Precautionary Statement
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
- P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P312-Call a POISON CENTER/doctor/if you feel unwell.