missing translation for 'onlineSavingsMsg'
Learn More

2-Methoxyethanol, CHROMASOLV™, for HPLC, ≥99.9%, Honeywell Riedel-de Haën™

Product Code. 15602850
Change view
Click to view available options
Quantity:
1 L
Packaging:
Glass Bottle
Unit Size:
1 L
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
15602850 1 L Glass Bottle 1 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15602850 Supplier Honeywell-Chromasolv Supplier No. 2704821L
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

CHROMASOLV™, for HPLC,≥99.9%

  • For HPLC and organic synthesis
  • High-purity, multipurpose solvents
  • High lot-to-lot consistency
TRUSTED_SUSTAINABILITY

Specifications

CAS 109-86-4
Absorbance max. 0.01 at 400 nm, max. 0.01 at 300 nm, max. 0.05 at 260 nm, max. 0.20 at 240 nm, max. 1.0 at 213 nm
CAS Max % 100.0000%
Melting Point -85°C (literature)
Density 0.965 g/mL at 25 °C(lit.)
Boiling Point 124°C
Flash Point 38°C
Packaging Glass Bottle
Molecular Formula C3H8O2
Linear Formula CH3OCH2CH2OH
MDL Number MFCD00002867
Quantity 1 L
UN Number UN1188
Beilstein 1731074
Synonym methyl cellosolve, ethanol, 2-methoxy, ethylene glycol monomethyl ether, methyl oxitol, 2-methoxy-1-ethanol, methoxyethanol, 3-oxa-1-butanol, egme, monomethyl glycol, dowanol em
InChI Key XNWFRZJHXBZDAG-UHFFFAOYSA-N
SMILES COCCO
IUPAC Name 2-methoxyethanol
Molecular Weight (g/mol) 76.095
PubChem CID 8019
ChEBI CHEBI:46790
Formula Weight 76.09
Percent Purity >99.9%
Grade HPLC
Chemical Name or Material 2-Methoxyethanol, CHROMASOLV™, for HPLC, ≥99.9%
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.