missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2-Methoxyethanol, ACS Reagent, ≥99.3%, Honeywell™
Click to view available options
Quantity:
500 mL
1 L
Packaging:
Glass Bottle
Unit Size:
1 L
500mL
Specifications
Specifications
| Melting Point | -85°C |
| Density | 0.965g/mL at 25°C |
| Boiling Point | 125°C |
| Explosion Limit | 0.245 |
| Flash Point | 40°C (104°F) |
| Ignition Point | 548°F |
| Infrared Spectrum | Conforms to structure |
| Odor | Characteristic |
| Packaging | Glass Bottle |
| Linear Formula | CH3OCH2CH2OH |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction