missing translation for 'onlineSavingsMsg'
Learn More

2-Methoxyethanol, 99%

Product Code. 11428057
Change view
Click to view available options
Quantity:
10,000 mL
2500 mL
500 mL
Unit Size:
10000mL
2500mL
500mL
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11428057 2500 mL 2500mL
11418057 500 mL 500mL
11438057 10,000 mL 10000mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11428057 Supplier Thermo Scientific Alfa Aesar Supplier No. A17459.0F
 Restricted Item
In Stock
Add to basket
This item is not returnable. View return policy

CAS: 109-86-4 | C3H8O2 | 76.095 g/mol

Solvent for ninhydrin color reagent2-Methoxy ethanol is used as a solvent for various purposes like varnishes, dyes, cleaning agents, epoxy coating, paints, anti-icing fuel additive in aviation and resins. It is involved in the preparation of Vaska’s complex viz. trans-chlorocarbonylbis(triphenylphosphine)iridium(I) and ruthenium hydridechlorocarbonyl tristriphenylphosphine. Further, it is also used in the synthesis of 2-aminothiazoles and Rho-associated kinase inhibitors.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Solvent for ninhydrin color reagent2-Methoxy ethanol is used as a solvent for various purposes like varnishes, dyes, cleaning agents, epoxy coating, paints, anti-icing fuel additive in aviation and resins. It is involved in the preparation of Vaska’s complex viz. trans-chlorocarbonylbis(triphenylphosphine)iridium(I) and ruthenium hydridechlorocarbonyl tristriphenylphosphine. Further, it is also used in the synthesis of 2-aminothiazoles and Rho-associated kinase inhibitors.

Solubility
Miscible with water, alcohol, ether, acetone, hydrocarbons, ketones, glycols and dimethylformamide.

Notes
Incompatible with strong bases, strong oxidizing agents, acids, bases, copper alloys and copper.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 109-86-4
Molecular Formula C3H8O2
Molecular Weight (g/mol) 76.095
MDL Number MFCD00002867
InChI Key XNWFRZJHXBZDAG-UHFFFAOYSA-N
Synonym methyl cellosolve, ethanol, 2-methoxy, ethylene glycol monomethyl ether, methyl oxitol, 2-methoxy-1-ethanol, methoxyethanol, 3-oxa-1-butanol, egme, monomethyl glycol, dowanol em
PubChem CID 8019
ChEBI CHEBI:46790
IUPAC Name 2-methoxyethanol
SMILES COCCO

Specifications

Melting Point -85°C
Density 0.965
Boiling Point 124°C to 125°C
Flash Point 46°C (115°F)
Odor Characteristic
Linear Formula CH3OCH2CH2OH
Refractive Index 1.4025
Quantity 2500 mL
UN Number UN1188
Beilstein 1731074
Merck Index 14,6038
Solubility Information Miscible with water,alcohol,ether,acetone,hydrocarbons,ketones,glycols and dimethylformamide.
Formula Weight 76.1
Percent Purity 99%
Chemical Name or Material 2-Methoxyethanol
Show More Show Less
compliance-icons compliance-icons compliance-icons
Product Identifier
  • 2-Methoxyethanol
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 4
  • Flammable liquid Category 3
  • Reproductive toxicity Category 1B
  • Specific target organ toxicity after repeated exposure Category 2
  • Specific target organ toxicity after single exposure Category 1
Hazard Statement
  • H226-Flammable liquid and vapour.
  • H302-Harmful if swallowed.
  • H312-Harmful in contact with skin.
  • H332-Harmful if inhaled.
  • H360FD-May damage fertility. May damage the unborn child.
  • H370-Causes damage to organs.
  • H373-May cause damage to organs.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.
Supplemental information
  • MIXTURE LIST-Contain: 2-Methoxyethanol

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.