missing translation for 'onlineSavingsMsg'
Learn More

2-Methoxyethanol, 99+%, extra pure

Product Code. 10183860 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
1 L
2.5 L
25 L
Packaging:
Glass Bottle
Metal drum
Unit Size:
1 L
2.5 L
25 L
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10183860 25 L Metal drum 25 L
10285660 1 L Glass Bottle 1 L
10317650 2.5 L Glass Bottle 2.5 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10183860 Supplier Thermo Scientific Acros Supplier No. 180790250
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

2-Methoxyethanol, 99+%, C3H8O2, CAS Number-109-86-4, 2-methoxy-1-ethanol, 3-oxa-1-butanol, dowanol em, egme, ethanol, 2-methoxy, ethylene glycol monomethyl ether, methoxyethanol, methyl cellosolve, methyl oxitol, monomethyl glycol, 2.5L, 99% min. (GC), 01, 467, 124.0 deg.C, 100.0, 99.0, CHEBI:46790 | CAS: 109-86-4 | C3H8O2 | 76.09 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 109-86-4
Molecular Formula C3H8O2
Molecular Weight (g/mol) 76.09
MDL Number MFCD00002867
InChI Key XNWFRZJHXBZDAG-UHFFFAOYSA-N
Synonym methyl cellosolve, ethanol, 2-methoxy, ethylene glycol monomethyl ether, methyl oxitol, 2-methoxy-1-ethanol, methoxyethanol, 3-oxa-1-butanol, egme, monomethyl glycol, dowanol em
PubChem CID 8019
ChEBI CHEBI:46790
IUPAC Name 2-methoxyethanol
SMILES COCCO

Specifications

CAS Min % 99.0
CAS Max % 100.0
Melting Point -85.0°C
Density 0.9600g/mL
Boiling Point 124.0°C
Flash Point 38°C
Infrared Spectrum Authentic
Assay Percent Range 99% min. (GC)
Packaging Metal drum
Linear Formula CH3OCH2CH2OH
Refractive Index 1.402
Quantity 25 L
Beilstein 01, 467
Residue after Evaporation 0.005% max.
Merck Index 15, 6063
Specific Gravity 0.96
Solubility Information Solubility in water: soluble
Viscosity 1.7 mPa.s (20°C)
Formula Weight 76.09
Percent Purity 99+%
Grade Extra Pure
Physical Form Liquid
Chemical Name or Material 2-Methoxyethanol, 99+%
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.