missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2-Methoxyestradiol, Mikromol™
Click to view available options
Quantity:
25 mg
100 mg
4 x 25 mg
Unit Size:
100mg
25mg
Description
2-Methoxyestradiol
Specifications
Specifications
| Chemical Name or Material | 2-Methoxyestradiol |
| CAS | 362-07-2 |
| Analyte or Component Names | 2-Methoxyestradiol |
| Shipping Condition | Room Temperature |
| Molecular Formula | C19 H26 O3 |
| Certifications/Compliance | ISO 17025 |
| Synonym | Estradiol, 2-methoxy- (6CI), 2-Hydroxyestradiol 2-methyl ether, 2-Methoxy-E2, 2-Methoxyestradiol, 2ME2, NSC 659853, Panzem, (17beta)-2-Methoxyestra-1,3,5(10)-triene-3,17-diol, Estra-1,3,5(10)-triene-3,17beta-diol, 2-methoxy- (7CI,8CI), 2-Methoxyestra-1,3,5(10)-triene-3,17beta-diol |
| InChI Formula | InChI=1S/C19H26O3/c1-19-8-7-12-13(15(19)5-6-18(19)21)4-3-11-9-16(20)17(22-2)10-14(11)12/h9-10,12-13,15,18,20-21H,3-8H2,1-2H3/t12-,13+,15-,18-,19-/m0/s1 |
| SMILES | COc1cc2[C@H]3CC[C@]4(C)[C@@H](O)CC[C@H]4[C@@H]3CCc2cc1O |
| IUPAC Name | (8R,9S,13S,14S,17S)-2-methoxy-13-methyl-6,7,8,9,11,12,14,15,16,17-decahydrocyclopenta[a]phenanthrene-3,17-diol |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction