missing translation for 'onlineSavingsMsg'
Learn More

2-Methoxycrotonic acid, 98%, Thermo Scientific™

Catalog Number 11314186
Click to view available options
Quantity:
5 g
25 g
This item is not returnable. View return policy
38588-37-3
C5H8O3
116.116
MFCD03426027
NVPNCVRJVRLJMU-UHFFFAOYSA-N
2-methoxycrotonic acid, acmc-20anrn, 2-butenoic acid, 2-methoxy-, z
53411933
2-methoxybut-2-enoic acid
CC=C(C(=O)O)OC
This item is not returnable. View return policy
38588-37-3
C5H8O3
116.116
MFCD03426027
NVPNCVRJVRLJMU-UHFFFAOYSA-N
2-methoxycrotonic acid, acmc-20anrn, 2-butenoic acid, 2-methoxy-, z
53411933
2-methoxybut-2-enoic acid
CC=C(C(=O)O)OC

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY
Melting Point 57°C to 58°C
Boiling Point 117°C (16 mmHg)
Quantity 5 g
UN Number UN3261
Beilstein 1745123
Formula Weight 116.12
Percent Purity 98%
Chemical Name or Material 2-Methoxycrotonic acid
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'