missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2-Methoxycarbonylphenylboronic acid, TRC
Click to view available options
Quantity:
100 mg
500 mg
Unit Size:
100mg
500mg
Specifications
Specifications
| Chemical Name or Material | 2-Methoxycarbonylphenylboronic acid |
| CAS | 374538-03-1 |
| Molecular Formula | C8H9BO4 |
| Quantity | 100 mg |
| InChI Formula | InChI=1S/C8H9BO4/c1-13-8(10)6-4-2-3-5-7(6)9(11)12/h2-5,11-12H,1H3 |
| SMILES | COC(=O)c1ccccc1B(O)O |
| IUPAC Name | (2-methoxycarbonylphenyl)boronic acid |
| Molecular Weight (g/mol) | 179.97 |
| Formula Weight | 180.0594 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction