missing translation for 'onlineSavingsMsg'
Learn More

2-(Methoxycarbonyl)phenyl isothiocyanate, 98%, Thermo Scientific™

Product Code. 11445536
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11445536 25 g 25g
11435536 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11445536 Supplier Thermo Scientific Alfa Aesar Supplier No. B20693.14

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 16024-82-1
Molecular Formula C9H7NO2S
Molecular Weight (g/mol) 193.22
MDL Number MFCD00041062
InChI Key UNXVHBOJSCWVCD-UHFFFAOYSA-N
PubChem CID 85243
IUPAC Name methyl 2-isothiocyanatobenzoate
SMILES COC(=O)C1=CC=CC=C1N=C=S

Specifications

Density 1.24
Boiling Point 149°C to 150°C (17 mmHg)
Refractive Index 1.6355
Quantity 25 g
UN Number UN2922
Beilstein 2720655
Sensitivity Moisture sensitive
Synonym 2-isothiocyanato-benzoic acid methyl ester, 2-methoxycarbonylphenyl isothiocyanate, 2-methoxycarbonyl phenyl isothiocyanate, benzoic acid,2-isothiocyanato-, methyl ester, benzoic acid, 2-isothiocyanato-, methyl ester, acmc-209dkr, methyl2-isothiocyanatobenzoate, methyl 2-isothiocyanato benzoate, methyl 2-isothiocyanatobenzenecarboxylate, 2-isothiocyanatobenzoic acid methyl ester
Formula Weight 193.23
Percent Purity 98%
Chemical Name or Material 2-(Methoxycarbonyl)phenyl isothiocyanate
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.