missing translation for 'onlineSavingsMsg'
Learn More

2-(Methoxycarbonyl)benzenesulfonamide, 98%

Product Code. 11480044
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
Unit Size:
100g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11480044 100 g 100g
11460044 5 g 5g
11470044 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11480044 Supplier Thermo Scientific Alfa Aesar Supplier No. A13553.22
 Restricted Item
In Stock
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 57683-71-3 | C8H9NO4S | 215.22 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 57683-71-3
Molecular Formula C8H9NO4S
Molecular Weight (g/mol) 215.22
MDL Number MFCD00009808
InChI Key VSOOBQALJVLTBH-UHFFFAOYSA-N
Synonym methyl 2-aminosulfonyl benzoate, 2-carbomethoxybenzenesulfonamide, 2-sulfamoylbenzoic acid methyl ester, 2-methoxycarbonyl benzene sulfonamide, methyl o-sulphamoylbenzoate, benzoic acid, 2-aminosulfonyl-, methyl ester, benzoic acid, 2-sulfamoyl-, methyl ester, 2-methoxycarbonylphenylsulfonamide, 2-methoxycarbonylbenzenesulfonamide, 2-methoxycarbonyl benzenesulphonamide
PubChem CID 42546
ChEBI CHEBI:83512
IUPAC Name methyl 2-sulfamoylbenzoate
SMILES COC(=O)C1=CC=CC=C1S(N)(=O)=O

Specifications

Melting Point 123°C to 125°C
Quantity 100 g
Formula Weight 215.23
Percent Purity 98%
Chemical Name or Material 2-(Methoxycarbonyl)benzenesulfonamide

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.