missing translation for 'onlineSavingsMsg'
Learn More

2-Methoxybiphenyl, 98+%

Product Code. 15458789
Change view
Click to view available options
Quantity:
25 g
Unit Size:
25g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15458789 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15458789 Supplier Thermo Scientific Alfa Aesar Supplier No. L01063.14

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

CAS: 86-26-0 | C13H12O | 184.24 g/mol

2-Methoxybiphenyl is used as chemical and organic intermediates.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-Methoxybiphenyl is used as chemical and organic intermediates.

Solubility
Soluble in toluene. Insoluble in water.

Notes
Incompatible with oxidizing agents.
TRUSTED_SUSTAINABILITY

Specifications

CAS 86-26-0
Melting Point ∼30°C
Density 1.023
Boiling Point 273°C to 274°C
Flash Point >110°C (230°F)
Molecular Formula C13H12O
Refractive Index 1.607
MDL Number MFCD00008367
Quantity 25 g
Beilstein 2045749
Synonym 2-methoxybiphenyl, 2-phenylanisole, 1,1'-biphenyl, 2-methoxy, o-phenylanisole, anisole, o-phenyl, methyl diphenyl ether, o-phenyl anisole, biphenyl-2-yl methyl ether, 2-methoxy-1,1'-biphenyl, 2-methoxy-biphenylate
Solubility Information Soluble in toluene. Insoluble in water.
InChI Key NLWCWEGVNJVLAX-UHFFFAOYSA-N
SMILES COC1=CC=CC=C1C1=CC=CC=C1
IUPAC Name 1-methoxy-2-phenylbenzene
Molecular Weight (g/mol) 184.24
PubChem CID 6835
Formula Weight 184.24
Percent Purity ≥98%
Chemical Name or Material 2-Methoxybiphenyl
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.