missing translation for 'onlineSavingsMsg'
Learn More

2'-Methoxybiphenyl-2-carboxaldehyde, 97+%, Thermo Scientific™

Product Code. 15479077
Change view
Click to view available options
Quantity:
250 mg
1 g
5 g
Unit Size:
1g
250mg
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15479077 250 mg 250mg
15459077 1 g 1g
15469077 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 15479077 Supplier Thermo Scientific Alfa Aesar Supplier No. H52384.MD
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 93465-26-0
Molecular Formula C14H12O2
Molecular Weight (g/mol) 212.248
MDL Number MFCD03424639
InChI Key ZYNNXRUYVVQLOW-UHFFFAOYSA-N
Synonym 2'-methoxy-biphenyl-2-carbaldehyde, 2-2-methoxyphenyl benzaldehyde, 2'-methoxybiphenyl-2-carbaldehyde, 2'-methoxy 1,1'-biphenyl-2-carbaldehyde, 2'-methoxy-1,1'-biphenyl-2-carbaldehyde, 2'-methoxybiphenyl-2-carboxaldehyde, 2'-methoxy 1,1'-biphenyl-2-carboxaldehyde, 2-methoxy-biphenyl-2-carbaldehyde, 2'-methoxy-1,1'-biphenyl-2-carboxaldehyde, 2'-methoxy 1,1'-biphenyl-2-carbaldehyde #
PubChem CID 602528
IUPAC Name 2-(2-methoxyphenyl)benzaldehyde
SMILES COC1=CC=CC=C1C2=CC=CC=C2C=O

Specifikationer

Quantity 250 mg
Sensitivity Air Sensitive
Formula Weight 212.24
Percent Purity ≥97%
Chemical Name or Material 2'-Methoxybiphenyl-2-carboxaldehyde
Produkttitel
Välj ett problem

Genom att klicka på Skicka bekräftar du att du kan bli kontaktad av Fisher Scientific angående feedbacken du har lämnat i detta formulär. Vi kommer inte att dela din information för andra ändamål. All kontaktinformation som tillhandahålls ska också underhållas i enlighet med vår Sekretesspolicy.