missing translation for 'onlineSavingsMsg'
Learn More

2-Methoxybenzylamine, 98+%

Product Code. 11377645
Change view
Click to view available options
Quantity:
25 g
100 g
Unit Size:
100g
25g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11377645 100 g 100g
11367645 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11377645 Supplier Thermo Scientific Alfa Aesar Supplier No. L02273.22
 Restricted Item
In Stock
Add to basket
This item is not returnable. View return policy

CAS: 6850-57-3 | C8H11NO | 137.182 g/mol

2-Methoxybenzylamine is used in the preparation of 6-substituted purines. It is also is used in the preparation of GABAA receptor ligands as analgesic agents.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-Methoxybenzylamine is used in the preparation of 6-substituted purines. It is also is used in the preparation of GABAA receptor ligands as analgesic agents.

Solubility
Soluble in water.

Notes
Keep container tightly sealed. Store in cool, dry conditions in well sealed containers. Incompatible with oxidizing agents. Store under dry inert gas. It is sensitive to air.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 6850-57-3
Molecular Formula C8H11NO
Molecular Weight (g/mol) 137.182
MDL Number MFCD00008110
InChI Key PXJACNDVRNAFHD-UHFFFAOYSA-N
Synonym 2-methoxybenzylamine, 2-methoxyphenyl methanamine, o-methoxybenzylamine, benzenemethanamine, 2-methoxy, 2-methoxy-benzylamine, 2-methoxyphenyl methylamine, 1-2-methoxyphenyl methanamine, 2-methyoxybenzylamine, 2-methoxy benzylamine, 2-methoxybenzyl amine
PubChem CID 81292
IUPAC Name (2-methoxyphenyl)methanamine
SMILES COC1=CC=CC=C1CN

Specifications

Density 1.057
Boiling Point 228°C to 229°C
Flash Point 100°C (212°F)
Refractive Index 1.548
Quantity 100 g
UN Number UN2735
Beilstein 1100448
Sensitivity Air Sensitive
Solubility Information Soluble in water.
Formula Weight 137.18
Percent Purity ≥98%
Chemical Name or Material 2-Methoxybenzylamine
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.