missing translation for 'onlineSavingsMsg'
Learn More

2-Methoxybenzyl alcohol, 99%

Product Code. 11487886
Change view
Click to view available options
Quantity:
25 g
100 g
Unit Size:
100g
25g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11487886 100 g 100g
11477886 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11487886 Supplier Thermo Scientific Alfa Aesar Supplier No. A14016.22
 Restricted Item
Estimated Shipment: 26-11-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 612-16-8 | C8H10O2 | 138.166 g/mol

It is an important raw material and intermediate used in Organic Synthesis, Pharmaceuticals, Agrochemicals.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is an important raw material and intermediate used in Organic Synthesis, Pharmaceuticals, Agrochemicals.

Solubility
Not miscible or difficult to mix in water. Soluble in ethanol.

Notes
Store in a cool, dry, well-ventilated area. Store away from oxidizing agent, strong acids.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 612-16-8
Molecular Formula C8H10O2
Molecular Weight (g/mol) 138.166
MDL Number MFCD00004611
InChI Key WYLYBQSHRJMURN-UHFFFAOYSA-N
Synonym 2-methoxybenzyl alcohol, 2-methoxyphenyl methanol, o-methoxybenzyl alcohol, benzenemethanol, 2-methoxy, o-anisyl alcohol, 2-methoxybenzylalcohol, 2-methoxybenzenemethanol, unii-457nq8ngcg, 457nq8ngcg, 2-methoxyphenyl methan-1-ol
PubChem CID 69154
IUPAC Name (2-methoxyphenyl)methanol
SMILES COC1=CC=CC=C1CO

Specifications

Density 1.12
Boiling Point 248°C to 250°C
Flash Point >110°C (230°F)
Refractive Index 1.5475
Quantity 100 g
Beilstein 2043673
Solubility Information Not miscible or difficult to mix in water. Soluble in ethanol.
Formula Weight 138.17
Percent Purity 99%
Chemical Name or Material 2-Methoxybenzyl alcohol

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.