missing translation for 'onlineSavingsMsg'
Learn More

2-Methoxybenzophenone, 98%

Product Code. 11450987
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11450987 5 g 5g
11440987 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11450987 Supplier Thermo Scientific Alfa Aesar Supplier No. B23376.06
 Restricted Item
In Stock
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 2553-04-0 | C14H12O2 | 212.248 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2553-04-0
Molecular Formula C14H12O2
Molecular Weight (g/mol) 212.248
MDL Number MFCD00017163
InChI Key CSUUDNFYSFENAE-UHFFFAOYSA-N
Synonym 2-methoxybenzophenone, o-methoxybenzophenone, 2-methoxyphenyl phenyl methanone, benzophenone, 2-methoxy, methanone, 2-methoxyphenyl phenyl, 2-methoxyphenyl phenyl ketone, methoxybenzophenone, acmc-1cotk, maybridge1_005821, 2-methoxyphenyl-phenylmethanone
PubChem CID 75702
IUPAC Name (2-methoxyphenyl)-phenylmethanone
SMILES COC1=CC=CC=C1C(=O)C2=CC=CC=C2

Specifications

Melting Point 36°C to 38°C
Boiling Point 149°C to 150°C (1 mmHg)
Quantity 5 g
Beilstein 1873339
Formula Weight 212.25
Percent Purity 98%
Chemical Name or Material 2-Methoxybenzophenone

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.