missing translation for 'onlineSavingsMsg'
Learn More

2-Methoxy-N-methylbenzylamine, 97%, Thermo Scientific™

Catalog Number 15419988
Click to view available options
Quantity:
1 g
5 g
This item is not returnable. View return policy
6851-80-5
C9H13NO
151.209
MFCD00025755
JCCQJCOMFAJJCQ-UHFFFAOYSA-N
2-methoxy-n-methylbenzylamine, 1-2-methoxyphenyl-n-methylmethanamine, 2-methoxybenzyl methylamine, 2-methoxyphenyl methyl methyl amine, n-methyl-2-methoxybenzylamine, benzenemethanamine, 2-methoxy-n-methyl, n-methyl-o-methoxybenzylamine, acmc-20a7fv, ksc495a5l
522302
1-(2-methoxyphenyl)-N-methylmethanamine
CNCC1=CC=CC=C1OC
This item is not returnable. View return policy
6851-80-5
C9H13NO
151.209
MFCD00025755
JCCQJCOMFAJJCQ-UHFFFAOYSA-N
2-methoxy-n-methylbenzylamine, 1-2-methoxyphenyl-n-methylmethanamine, 2-methoxybenzyl methylamine, 2-methoxyphenyl methyl methyl amine, n-methyl-2-methoxybenzylamine, benzenemethanamine, 2-methoxy-n-methyl, n-methyl-o-methoxybenzylamine, acmc-20a7fv, ksc495a5l
522302
1-(2-methoxyphenyl)-N-methylmethanamine
CNCC1=CC=CC=C1OC

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY
Color Colorless
Density 1.015 g/mL
Boiling Point 234°C to 235°C
Flash Point 106°C (223°F)
Refractive Index 1.5235
Quantity 5 g
UN Number UN2735
Formula Weight 151.21
Percent Purity 97%
Physical Form Liquid
Chemical Name or Material 2-Methoxy-N-methylbenzylamine
Show More Show Less
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'