missing translation for 'onlineSavingsMsg'
Learn More

2-Methoxy-6-methylaniline, TRC

Product Code. 17944515
Change view
Click to view available options
Quantity:
100 mg
250 mg
1 g
Unit Size:
100mg
1g
250mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
17944515 100 mg 100mg
30136111 250 mg 250mg
30135781 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 17944515 Supplier Toronto Research Chemicals Supplier No. TRCM329148100MG
 Restricted Item
Estimated Shipment: 13-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 50868-73-0
Molecular Formula C8 H11 N O
Molecular Weight (g/mol) 137.179
Synonym 2-Methoxy-6-methylbenzenamine, 2-Methoxy-6-methylphenylamine, 2-Methyl-6-methoxyaniline
IUPAC Name 2-methoxy-6-methyl-aniline
SMILES COc1cccc(C)c1N

Specifications

Quantity 100 mg
InChI Formula InChI=1S/C8H11NO/c1-6-4-3-5-7(10-2)8(6)9/h3-5H,9H2,1-2H3
Formula Weight 137.0841
Chemical Name or Material 2-Methoxy-6-methylaniline
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.