missing translation for 'onlineSavingsMsg'
Learn More

2-Methoxy-5-(trifluoromethyl)benzoic acid, Maybridge

Product Code. 10893581
Change view
Click to view available options
Quantity:
5 g
Packaging:
Amber Glass Bottle
Unit Size:
5g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10893581 5 g Amber Glass Bottle 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10893581 Supplier Thermo Scientific Maybridge Supplier No. MAY00126DE

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Specifications

CAS 4876-10-2
Packaging Amber Glass Bottle
Molecular Formula C10H8BrNO
Quantity 5 g
Synonym 4-bromomethyl-1,2-dihydroquinoline-2-one, 4-bromomethyl quinolin-2 1h-one, 4-bromomethyl-2 1h-quinolinone, 4-bromomethyl-1,2-dihydroquinolin-2-one, 4-bromomethyl-1h-quinolin-2-one, 2 1h-quinolinone, 4-bromomethyl, 4-bromomethyl-2-quinolinone, 2-hydroxy-4-bromo methyl quinoline, 4-bromomethyl hydroquinolin-2-one, 4-bromomethylquinolinon
InChI Key BBAHJCUCNVVEQU-UHFFFAOYSA-N
SMILES C1=CC=C2C(=C1)C(=CC(=O)N2)CBr
IUPAC Name 4-(bromomethyl)-1H-quinolin-2-one
Molecular Weight (g/mol) 238.08
PubChem CID 268296
Formula Weight 220.15
Percent Purity 97%
Chemical Name or Material 2-methoxy-5-(trifluoromethyl)benzoic acid
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.