missing translation for 'onlineSavingsMsg'
Learn More

2-Methoxy-5-(trifluoromethyl)benzaldehyde, 98%, Thermo Scientific Chemicals

Product Code. p-7028729
Change view
Click to view available options
Quantity:
250 mg
1 g
Unit Size:
1g
250mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15457555 250 mg 250mg
11322139 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15457555 Supplier Thermo Scientific Alfa Aesar Supplier No. H26135.MD

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 146539-83-5 | Molecular Formula: C9H7F3O2 | Molecular Weight: 204.15 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 146539-83-5
Molecular Formula C9H7F3O2
Molecular Weight (g/mol) 204.15
MDL Number MFCD03788528
InChI Key WFRURKYDETYZGF-UHFFFAOYSA-N
Synonym 2-methoxy-5-trifluoromethyl benzaldehyde, 2-methoxy-5-trifluoromethyl-benzaldehyde, benzaldehyde, 2-methoxy-5-trifluoromethyl, 5-trifluoromethyl-2-methoxybenzaldehyde, pubchem8509, acmc-209cx7, ksc496k3j, benzaldehyde,2-methoxy-5-trifluoromethyl
PubChem CID 2779272
IUPAC Name 2-methoxy-5-(trifluoromethyl)benzaldehyde
SMILES COC1=CC=C(C=C1C=O)C(F)(F)F

Specifications

Melting Point 85°C to 89°C
Quantity 250 mg
Sensitivity Air Sensitive
Formula Weight 204.15
Percent Purity 98%
Chemical Name or Material 2-Methoxy-5-(trifluoromethyl)benzaldehyde

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.