missing translation for 'onlineSavingsMsg'
Learn More

2-Methoxy-5-(trifluoromethoxy)benzoic acid, 96%, Thermo Scientific™

Product Code. p-7028699
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11322079 1 g 1g
15493377 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11322079 Supplier Thermo Scientific Alfa Aesar Supplier No. H26103.03

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 191604-88-3
Molecular Formula C9H7F3O4
Molecular Weight (g/mol) 236.146
MDL Number MFCD01862055
InChI Key HURBWIHJHDFCGU-UHFFFAOYSA-N
Synonym 2-methoxy-5-trifluoromethoxy benzoic acid, 3-trifluoromethoxy-6-methoxybenzoic acid, 2-methoxy-5-trifluoromethoxy-benzoic acid, benzoic acid,2-methoxy-5-trifluoromethoxy, pubchem4509, acmc-1bzxk, rarechem al bo 1035, benzoic acid, 2-methoxy-5-trifluoromethoxy, 2-methoxy-5-trifluoromethoxy benzoicacid
PubChem CID 2775286
IUPAC Name 2-methoxy-5-(trifluoromethoxy)benzoic acid
SMILES COC1=C(C=C(C=C1)OC(F)(F)F)C(=O)O

Specifications

Melting Point 63°C to 68°C
Quantity 1 g
Sensitivity Air Sensitive
Formula Weight 236.15
Percent Purity 96%
Chemical Name or Material 2-Methoxy-5-(trifluoromethoxy)benzoic acid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.