missing translation for 'onlineSavingsMsg'
Learn More

2-Methoxy-5-sulfamoylbenzoic Acid, TRC

Product Code. 30473597
Change view
Click to view available options
Quantity:
1 g
10 g
50 g
Unit Size:
10g
1g
50g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30473597 1 g 1g
30484144 50 g 50g
30466685 10 g 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30473597 Supplier Toronto Research Chemicals Supplier No. TRCM3266401G
 Restricted Item
Estimated Shipment: 25-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 22117-85-7
Molecular Formula C8 H9 N O5 S
Molecular Weight (g/mol) 231.23
Synonym 2-Methoxy-5-sulphamoylbenzoic Acid, Sulpiride Imp. D (EP)
IUPAC Name 2-methoxy-5-sulfamoylbenzoic acid
SMILES COc1ccc(cc1C(=O)O)S(=O)(=O)N

Specifications

Quantity 1 g
InChI Formula InChI=1S/C8H9NO5S/c1-14-7-3-2-5(15(9,12)13)4-6(7)8(10)11/h2-4H,1H3,(H,10,11)(H2,9,12,13)
Formula Weight 231.0201
Chemical Name or Material 2-Methoxy-5-sulfamoylbenzoic acid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.