missing translation for 'onlineSavingsMsg'
Learn More

2-Methoxy-5-nitropyridine, 97%, Thermo Scientific™

Product Code. 10081280 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
25 g
100 g
Unit Size:
100g
25g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
10081280 100 g 100g
10436940 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10081280 Supplier Thermo Scientific Acros Supplier No. 125991000
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Chemical Identifiers

CAS 5446-92-4
Molecular Formula C6H6N2O3
Molecular Weight (g/mol) 154.13
MDL Number MFCD00006263
InChI Key WUPLOZFIOAEYMG-UHFFFAOYSA-N
Synonym 2-methoxy-5-nitro-pyridine, 5-nitro-2-methoxypyridine, 6-methoxy-3-nitropyridine, pyridine, 2-methoxy-5-nitro, methoxy 2--5-nitropyridine, 2-methoxy-5-nitropyridin, pubchem1207, acmc-1art3, 2-methoxy-5-nitro pyridine, 5-nitro-2-methoxy pyridine
PubChem CID 79525
IUPAC Name 2-methoxy-5-nitropyridine
SMILES COC1=CC=C(C=N1)[N+]([O-])=O

Specifications

CAS Min % 96.0
CAS Max % 100.0
Melting Point 108.0°C to 111.0°C
Color Colorless to Yellow
Density 0.45g/mL
Flash Point 130°C
Infrared Spectrum Authentic
Assay Percent Range 96% min. (GC)
Packaging Glass bottle
Linear Formula CH3O(C5H3N)NO2
Quantity 100 g
Beilstein 21, II, 33
Solubility Information Solubility in water: insoluble. Other solubilities: soluble in chlorinated hydrocarbons,,insoluble in hexane
Formula Weight 154.12
Percent Purity 97%
Physical Form Liquid
Chemical Name or Material 2-Methoxy-5-nitropyridine, 97%
Show More Show Less
compliance-icons
Product Identifier
  • 2-Methoxy-5-nitropyridine
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.