missing translation for 'onlineSavingsMsg'
Learn More

2-Methoxy-5-methylphenyl isothiocyanate, 97%, Thermo Scientific™

Product Code. 10387901 Shop All Thermo Scientific Products
Click to view available options
Quantity:
5 g
Packaging:
Glass bottle
Unit Size:
5g
This item is not returnable. View return policy
Regulatory: This product contains a substance listed in Annex XVII to REACH Regulation (EC) No 1907/2006, all customers must complete a declaration before delivery.

Product Code. 10387901

Brand: Thermo Scientific 333020050

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy
Regulatory: This product contains a substance listed in Annex XVII to REACH Regulation (EC) No 1907/2006, all customers must complete a declaration before delivery.

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Specifications

CAS 190774-56-2
CAS Min % 96.0
CAS Max % 100.0
Color Yellow
Density 1.1400g/mL
Boiling Point 130°C (0.5 mmHg)
Flash Point >110°C
Infrared Spectrum Authentic
Assay Percent Range 96% min. (GC)
Packaging Glass bottle
Molecular Formula C9H9NOS
Refractive Index 1.6290 to 1.631
MDL Number MFCD00041083
Quantity 5 g
Specific Gravity 1.14
Synonym 2-methoxy-5-methylphenyl isothiocyanate, 2-methoxy-5-methylphenylisothiocyanate, benzene,2-isothiocyanato-1-methoxy-4-methyl, acmc-20anrv, 2-methoxy-5-methylbenzenisothiocyanate, 2-isothiocyanato-1-methoxy-4-methyl-benzene, #, benzene, 2-isothiocyanato-1-methoxy-4-methyl-9ci
Solubility Information Solubility in water: insoluble
InChI Key CYKFAZDMLSXWKZ-UHFFFAOYSA-N
SMILES CC1=CC(=C(C=C1)OC)N=C=S
IUPAC Name 2-isothiocyanato-1-methoxy-4-methylbenzene
Molecular Weight (g/mol) 179.2
PubChem CID 601672
Formula Weight 179.2
Percent Purity 97%
Physical Form Liquid
Chemical Name or Material 2-Methoxy-5-methylphenyl isothiocyanate, 97%
Show More Show Less
compliance-icons compliance-icons compliance-icons
Product Identifier
  • 2-Methoxy-5-methylphenyl isothiocyanate
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 3
  • Serious eye damage/eye irritation Category 1
  • Respiratory sensitiser Category 1
  • Skin corrosion/irritation Category 1 B
  • Skin sensitiser Category 1
Hazard Statement
  • H317-May cause an allergic skin reaction.
  • H318-Causes serious eye damage.
  • H334-May cause allergy or asthma symptoms or breathing difficulties if inhaled.
  • H314-Causes severe skin burns and eye damage.
  • H311-Toxic in contact with skin.
  • H331-Toxic if inhaled.
  • H301-Toxic if swallowed.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.
  • P342+P311-If experiencing respiratory symptoms: Call a POISON CENTER/doctor/ .
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.