missing translation for 'onlineSavingsMsg'
Learn More

2-Methoxy-5-methylaniline, 95%

Product Code. p-3778986 Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
5 g
25 g
100 g
500 g
Packaging:
Glass bottle
Unit Size:
100g
25g
500g
5g
This item is not returnable. View return policy

Product Code. 10594281

Brand: Thermo Scientific Acros 332080050

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

CAS: 120-71-8 | C8H11NO | 137.18 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 120-71-8
Molecular Formula C8H11NO
Molecular Weight (g/mol) 137.18
MDL Number MFCD00007815
InChI Key WXWCDTXEKCVRRO-UHFFFAOYSA-N
Synonym p-cresidine, cresidine, 5-methyl-o-anisidine, para-cresidine, benzenamine, 2-methoxy-5-methyl, krezidin, 2-amino-4-methylanisole, 3-amino-4-methoxytoluene, 4-methoxy-m-toluidine, 6-methoxy-m-toluidine
PubChem CID 8445
ChEBI CHEBI:82307
IUPAC Name 2-methoxy-5-methylaniline
SMILES CC1=CC(=C(C=C1)OC)N

Specifications

CAS Min % 94.0
CAS Max % 100.0
Melting Point 49.0°C to 55.0°C
Color Brown to Black
Density 111g/mL
Boiling Point 235.0°C
Flash Point 111°C
Infrared Spectrum Authentic
Assay Percent Range 94% min. (GC)
Packaging Glass bottle
Linear Formula CH3OC6H3(CH3)NH2
Quantity 5 g
Beilstein 13, II, 338
Solubility Information Solubility in water: insoluble in water. Other solubilities: soluble in most common organic solvents
Formula Weight 137.18
Percent Purity 95%
Physical Form Crystalline Chunks
Chemical Name or Material 2-Methoxy-5-methylaniline
Show More Show Less

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.