missing translation for 'onlineSavingsMsg'
Learn More

2-Methoxy-5-methylaniline, 95%

Product Code. 10390114 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
500 g
Packaging:
Glass bottle
Unit Size:
100g
25g
500g
5g
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10390114 100 g Glass bottle 100g
10594281 5 g Glass bottle 5g
10183051 25 g Glass bottle 25g
10711175 500 g Glass bottle 500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 10390114 Supplier Thermo Scientific Acros Supplier No. 332081000
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

CAS: 120-71-8 | C8H11NO | 137.18 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 120-71-8
Molecular Formula C8H11NO
Molecular Weight (g/mol) 137.18
MDL Number MFCD00007815
InChI Key WXWCDTXEKCVRRO-UHFFFAOYSA-N
Synonym p-cresidine, cresidine, 5-methyl-o-anisidine, para-cresidine, benzenamine, 2-methoxy-5-methyl, krezidin, 2-amino-4-methylanisole, 3-amino-4-methoxytoluene, 4-methoxy-m-toluidine, 6-methoxy-m-toluidine
PubChem CID 8445
ChEBI CHEBI:82307
IUPAC Name 2-methoxy-5-methylaniline
SMILES CC1=CC(=C(C=C1)OC)N

Specifications

CAS Min % 94.0
CAS Max % 100.0
Melting Point 49.0°C to 55.0°C
Color Brown to Black
Density 111g/mL
Boiling Point 235.0°C
Flash Point 111°C
Infrared Spectrum Authentic
Assay Percent Range 94% min. (GC)
Packaging Glass bottle
Linear Formula CH3OC6H3(CH3)NH2
Quantity 100 g
Beilstein 13, II, 338
Solubility Information Solubility in water: insoluble in water. Other solubilities: soluble in most common organic solvents
Formula Weight 137.18
Percent Purity 95%
Physical Form Crystalline Chunks
Chemical Name or Material 2-Methoxy-5-methylaniline
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.