missing translation for 'onlineSavingsMsg'
Learn More

2-Methoxy-5-formylphenylboronic acid, 97%, Thermo Scientific™

Catalog Number 10340894
Click to view available options
Quantity:
5 g
Packaging:
Glass bottle
This item is not returnable. View return policy
127972-02-5
C8H9BO4
179.97
MFCD01319044
NKKNXLPHCRLBDY-UHFFFAOYSA-N
2-methoxy-5-formylphenylboronic acid, 5-formyl-2-methoxyphenyl boronic acid, 5-formyl-2-methoxybenzeneboronic acid, 5-formyl-2-methoxy-phenyl boronic acid, 2-methoxy-5-formylbenzeneboronic acid, 3-borono-4-methoxybenzaldehyde, 3-dihydroxyboranyl-4-methoxybenzaldehyde, boronic acid, 5-formyl-2-methoxyphenyl, pubchem7991, acmc-209bd6
2734367
(5-formyl-2-methoxyphenyl)boronic acid
COC1=CC=C(C=O)C=C1B(O)O
This item is not returnable. View return policy
127972-02-5
C8H9BO4
179.97
MFCD01319044
NKKNXLPHCRLBDY-UHFFFAOYSA-N
2-methoxy-5-formylphenylboronic acid, 5-formyl-2-methoxyphenyl boronic acid, 5-formyl-2-methoxybenzeneboronic acid, 5-formyl-2-methoxy-phenyl boronic acid, 2-methoxy-5-formylbenzeneboronic acid, 3-borono-4-methoxybenzaldehyde, 3-dihydroxyboranyl-4-methoxybenzaldehyde, boronic acid, 5-formyl-2-methoxyphenyl, pubchem7991, acmc-209bd6
2734367
(5-formyl-2-methoxyphenyl)boronic acid
COC1=CC=C(C=O)C=C1B(O)O

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY
CAS Min % 96.0
CAS Max % 100.0
Color Orange
Infrared Spectrum Authentic
Assay Percent Range 97%
Packaging Glass bottle
Quantity 5 g
Formula Weight 179.97
Percent Purity ≥96.0%
Physical Form Powder
Chemical Name or Material 2-Methoxy-5-formylphenylboronic acid
Show More Show Less
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'