missing translation for 'onlineSavingsMsg'
Learn More

2-Methoxy-5-[(2R)-2-[[(1R)-1-phenylethyl]amino]propyl]benzenesulfonamide, Mikromol

Product Code. 30461357
Change view
Click to view available options
Quantity:
25 mg
Unit Size:
25mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30461357 25 mg 25mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30461357 Supplier Mikromol Supplier No. MM0938.170025

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Discover Mikromol- your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.

2-Methoxy-5-[(2R)-2-[[(1R)-1-phenylethyl]amino]propyl]benzenesulfonamide, Mikromol
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 2-Methoxy-5-[(2R)-2-[((1R)-1-phenylethyl)amino]propyl]benzenesulfonamide
CAS 121565-95-5
Analyte or Component Names 2-Methoxy-5-[(2R)-2-[((1R)-1-phenylethyl)amino]propyl]benzenesulfonamide
Shipping Condition Room Temperature
Molecular Formula C18 H24 N2 O3 S
Certifications/Compliance ISO 9001
Synonym Benzenesulfonamide, 2-methoxy-5-[(2R)-2-[[(1R)-1-phenylethyl]amino]propyl]-, 2-Methoxy-5-[(2R)-2-[[(1R)-1-phenylethyl]amino]propyl]benzenesulfonamide
InChI Formula InChI=1S/C18H24N2O3S/c1-13(20-14(2)16-7-5-4-6-8-16)11-15-9-10-17(23-3)18(12-15)24(19,21)22/h4-10,12-14,20H,11H2,1-3H3,(H2,19,21,22)/t13-,14-/m1/s1
Solution Type Neat
SMILES COc1ccc(C[C@@H](C)N[C@H](C)c2ccccc2)cc1S(=O)(=O)N
IUPAC Name 2-methoxy-5-[(2R)-2-[[(1R)-1-phenylethyl]amino]propyl]benzenesulfonamide
Molecular Weight (g/mol) 348.4598
Quantity 25 mg
Formula Weight 348.1508 g/mol
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.