missing translation for 'onlineSavingsMsg'
Learn More

2-Methoxy-4-nitroaniline, 99%

Product Code. 11491257
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
Unit Size:
100g
25g
500g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11491257 100 g 100g
11481257 25 g 25g
11401267 500 g 500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11491257 Supplier Thermo Scientific Alfa Aesar Supplier No. B24220.22
 Restricted Item
Estimated Shipment: 31-07-2026
Add to basket
This item is not returnable. View return policy

CAS: 97-52-9 | C7H8N2O3 | 168.15 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 97-52-9
Molecular Formula C7H8N2O3
Molecular Weight (g/mol) 168.15
MDL Number MFCD00007363
InChI Key GVBHRNIWBGTNQA-UHFFFAOYSA-N
Synonym fast red b base, benzenamine, 2-methoxy-4-nitro, 4-nitro-o-anisidine, fast red base b, fast red b, red b base, diazo fast red b, 2-amino-5-nitroanisole, azoamine pink o, fast red 5na base
PubChem CID 7337
IUPAC Name 2-methoxy-4-nitroaniline
SMILES COC1=CC(=CC=C1N)[N+]([O-])=O

Specifications

Melting Point 138°C to 142°C
Density 1.211
Quantity 100 g
Beilstein 879619
Formula Weight 168.15
Percent Purity 99%
Chemical Name or Material 2-Methoxy-4-nitroaniline

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.