missing translation for 'onlineSavingsMsg'
Learn More

2-Methoxy-4-methyl-5-nitropyridine, TRC

Product Code. 30477204
Change view
Click to view available options
Quantity:
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30477204 100 mg 100mg
30466619 50 mg 50mg
30475963 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30477204 Supplier Toronto Research Chemicals Supplier No. TRCM269468100MG
 Restricted Item
Estimated Shipment: 30-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 6635-90-1
Molecular Formula C7H8N2O3
Molecular Weight (g/mol) 168.15
Synonym NSC 52457
SMILES COc1cc(C)c(cn1)[N+](=O)[O-]

Specifications

Quantity 100 mg
InChI Formula InChI=1S/C7H8N2O3/c1-5-3-7(12-2)8-4-6(5)9(10)11/h3-4H,1-2H3
Formula Weight 168.0535
Chemical Name or Material 2-Methoxy-4-methyl-5-nitropyridine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.