missing translation for 'onlineSavingsMsg'
Learn More

2-Methoxy-1-naphthaldehyde, 99%

Product Code. p-7025138
Click to view available options
Quantity:
5 g
25 g
100 g
Unit Size:
100g
25g
5g
This item is not returnable. View return policy

Product Code. 11414477

Brand: Thermo Scientific Alfa Aesar B20483.06

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 5392-12-1 | C12H10O2 | 186.21 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 5392-12-1
MDL Number MFCD00004004

Specifications

Melting Point 81°C to 85°C
Boiling Point 204°C to 206°C (18 mmHg)
Molecular Formula C14H10O2
Quantity 5 g
Beilstein 1868308
Sensitivity Air Sensitive
Synonym 3-phenylphthalide, 3-phenyl phthalide, phthalide, 3-phenyl, 3-phenyl-2-benzofuran-1 3h-one, 3-phenyl-1 3h-isobenzofuranone, 3-phenyl-1,3-dihydro-2-benzofuran-1-one, 1 3h-isobenzofuranone, 3-phenyl, 3-phenyl-3-hydroisobenzofuran-1-one, phenylphthalid, acmc-1ak9n
InChI Key SQFMIHCARVMICF-UHFFFAOYNA-N
SMILES C1=CC=C(C=C1)C2C3=CC=CC=C3C(=O)O2
IUPAC Name 3-phenyl-3H-2-benzofuran-1-one
Molecular Weight (g/mol) 210.23
PubChem CID 95365
Formula Weight 186.21
Percent Purity 99%
Chemical Name or Material 2-Methoxy-1-naphthaldehyde
Show More Show Less

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.