missing translation for 'onlineSavingsMsg'
Learn More

2-(Methacryloyloxy)ethyltrimethylammonium chloride, 72% aq. soln., stab. with 150-200 ppm 4-methoxyphenol

Product Code. 11373117
missing translation for 'orderingAttributeHoverText'
Quantity:
100 g
500 g
missing translation for 'unitSize'
100g
500g
This item is not returnable. View return policy

Product Code. 11373117

missing translation for 'mfr': Thermo Scientific Alfa Aesar L13523.22

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 5039-78-1 | C9H18ClNO2 | 207.698 g/mol

2-(Methacryloyloxy)ethyltrimethylammonium chloride is a zirconium, aluminum and organic surface treated multipurpose rutile titanium dioxide pigment, it has narrow-size material distribution, good whiteness and high weather resistance. It is widely used in coating, plastics, paints, rubber and ink production, leather, cosmetics, soaps, plastics and in paper industry.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-(Methacryloyloxy)ethyltrimethylammonium chloride is a zirconium, aluminum and organic surface treated multipurpose rutile titanium dioxide pigment, it has narrow-size material distribution, good whiteness and high weather resistance. It is widely used in coating, plastics, paints, rubber and ink production, leather, cosmetics, soaps, plastics and in paper industry.

Solubility
It is soluble in water.

Notes
Keep container tightly closed in a dry and well-ventilated place. Keep away from oxidizing agents. Stable under recommended storage conditions.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 5039-78-1
Molecular Formula C9H18ClNO2
Molecular Weight (g/mol) 207.698
MDL Number MFCD00060097
InChI Key RRHXZLALVWBDKH-UHFFFAOYSA-M
Synonym polyquaternium-37, 2-methacryloyloxy-n,n,n-trimethylethanaminium chloride, unii-pp88r88k3o, methacrylatoethyl trimethyl ammonium chloride, 2-methacryloyloxy ethyl trimethylammonium chloride, 2-methacryloyloxy ethyl trimethylammonium, methacryloxyethyltrimethyl ammonium chloride, ethanaminium, n,n,n-trimethyl-2-2-methyl-1-oxo-2-propenyl oxy-, chloride
PubChem CID 78738
IUPAC Name trimethyl-[2-(2-methylprop-2-enoyloxy)ethyl]azanium;chloride
SMILES CC(=C)C(=O)OCC[N+](C)(C)C.[Cl-]

Specifications

Melting Point -25°C
Density 1.105
Boiling Point >100°C
Refractive Index 1.466
Quantity 100 g
Beilstein 8168395
Solubility Information It is soluble in water.
Formula Weight 207.7
Concentration or Composition (by Analyte or Components) 72% aq. soln.
Chemical Name or Material 2-(Methacryloyloxy)ethyltrimethylammonium chloride, stabilized with 150 to 200ppm 4-methoxyphenol
compliance-icons
Product Identifier
  • 2-(Methacryloyloxy)ethyltrimethylammoniumchloride, 72% in water
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.
  • P332+P313-If skin irritation occurs: Get medical advice/attention.
  • P337+P313-If eye irritation persists: Get medical advice/attention.
Supplemental information
  • MIXTURE LIST-Contains : 2-(Methacryloyloxy)ethyltrimethylammoniumchloride

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.