missing translation for 'onlineSavingsMsg'
Learn More

2-Mesitylmagnesium bromide, 1M solution in THF, AcroSeal™

Product Code. 10421820 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
100 mL
Packaging:
AcroSeal™ Glass Bottle
Unit Size:
100mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10421820 100 mL AcroSeal™ Glass Bottle 100mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10421820 Supplier Thermo Scientific Acros Supplier No. 210731000
  • ONLINE EXCLUSIVE PRICE
    £99.00
    £89.65 / 100mL
    Save £9.35
    9% Off
 Restricted Item
Estimated Shipment: 23-07-2026
Add to basket
This item is not returnable. View return policy

CAS: 2633-66-1 | C9H11BrMg | 223.40 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Specifications

CAS 2633-66-1, 109-99-9
Color Yellow to Brown
CAS Min % 21.12, 75.55
CAS Max % 24.45, 78.88
Physical Form Solution
Quantity 100 mL
Molecular Formula C9H11BrMg
Linear Formula (CH3)3C6H2MgBr
MDL Number MFCD00009663
Flash Point −17°C
Synonym 2-mesitylmagnesium bromide, mesitylmagnesium bromide, 2,4,6-trimethylphenylmagnesium bromide, 2-mesitylmagnesium bromide solution, mesmgbr, yxvsitsudrgill-uhfffaoysa-m, magnesium,bromo 2,4,6-trimethylphenyl, 2-mesitylmagnesium bromide solution, 1.0 m in thf, 2,4,6-trimethylphenylmagnesium bromide, 1m in 2-methf, 2-mesitylmagnesium bromide solution, 1.0 m in diethyl ether, grignard reagent
InChI Key VZMGGVXGYWKQPB-UHFFFAOYSA-M
SMILES CC1=CC([Mg]Br)=CC(C)=C1C
IUPAC Name bromo(3,4,5-trimethylphenyl)magnesium
Molecular Weight (g/mol) 223.40
PubChem CID 5098599
Formula Weight 223.39
Concentration or Composition (by Analyte or Components) 0.95 to 1.10M
Packaging AcroSeal™ Glass Bottle
Specific Gravity 1.005
Density 1.0050g/mL
Chemical Name or Material 2-Mesitylmagnesium bromide
Show More Show Less
compliance-icons compliance-icons compliance-icons compliance-icons
Product Identifier
  • 2-Mesitylmagnesium bromide, 1M solution in THF
Signal Word
  • Danger
Hazard Category
  • Carcinogenicity Category 2
  • Serious eye damage/eye irritation Category 1
  • Flammable liquid Category 2
  • Skin corrosion/irritation Category 1 B
  • Specific target organ toxicity Category 3
Hazard Statement
  • H318-Causes serious eye damage.
  • H314-Causes severe skin burns and eye damage.
  • H351-Suspected of causing cancer.
  • H335-May cause respiratory irritation.
  • H336-May cause drowsiness or dizziness.
  • H225-Highly flammable liquid and vapour.
  • EUH014-Reacts violently with water.
  • EUH019-May form explosive peroxides.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.