missing translation for 'onlineSavingsMsg'
Learn More

2-Mercaptopyridine N-Oxide Sodium Salt, TRC

Product Code. 30469925
Change view
Click to view available options
Quantity:
2.5 g
Unit Size:
2.5g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30469925 2.5 g 2.5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30469925 Supplier Toronto Research Chemicals Supplier No. TRCM2252602.5G
 Restricted Item
Estimated Shipment: 21-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 2-Mercaptopyridine N-Oxide Sodium Salt
CAS 3811-73-2
Molecular Formula C5H4NNaOS
Quantity 2.5 g
Synonym 2-Pyridinethiol 1-Oxide Sodium Salt (1:1), 2-Pyridinethiol 1-Oxide Sodium Deriv., 2-Pyridinethiol 1-Oxide Sodium Salt, Sodium (2-pyridylthio)-N-oxide, 1-Oxo-2-pyridinethiol Sodium Salt, 2-Mercaptopyridine 1-Oxide Sodium Salt, 2-Mercaptopyridine N-oxide Sodium Salt, 2-Mercaptopyridine Oxide Sodium Salt, 2-Pyridinethiol N-Oxide Sodium Salt, 2-Pyridylthiol-N-oxide Sodium Salt, Hokucide N 100, San-aibac Sodium Omadine, Sodium (2-pyridylthio-1-oxide), Sodium 2-mercaptopyridine 1-Oxide, Sodium 2-Mercaptopyridine N-Oxide, Sodium 2-Pyridinethiol 1-Oxide, Sodium 2-Pyridinethiol N-Oxide, Sodium 2-Pyridinethiolate 1-Oxide, Sodium 2-Pyridinethiolate N-Oxide, Sodium Pyridine-1-oxide-2-thiolate, Sodium Pyridine-2-thione-N-oxide, Sodium Pyrithione, Sodium (2-Pyridinylthio)-N-oxide, Thione (reagent), Tomicide S, Topcide 280
InChI Formula InChI=1S/C5H5NOS.Na/c7-6-4-2-1-3-5(6)8;/h1-4,8H;/q;+1/p-1
SMILES [Na+].[O-][n+]1ccccc1[S-]
IUPAC Name sodium;1-oxidopyridin-1-ium-2-thiolate
Molecular Weight (g/mol) 149.15
Formula Weight 148.9911
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.