missing translation for 'onlineSavingsMsg'
Learn More

2-Mercaptopyridine N-oxide sodium salt, anhydrous, 98%

Product Code. 11438897
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
Unit Size:
1g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11438897 1 g 1g
11448897 5 g 5g
11458897 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11438897 Supplier Thermo Scientific Alfa Aesar Supplier No. B20529.03
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 3811-73-2 | C5H4NNaOS | 149.143 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Suitable for battery materials development.

Applications
2-Mercaptopyridine N-oxide sodium salt is one of the active components in paint, sealants, shampoo, adhesive and aerosol due to its anti-microbial activity. In biochemistry studies, it is utilized to transport zinc into cells. Further, it is used to form bidentate oxothiolane chelates with transition metals. It acts as a stabilizer and viscosity building provider in weak basic or neutral medium.

Solubility
Soluble in water, ethanol, propylene glycol, polyethylene glycol and dimethyl sulfoxide. Insoluble in liquid paraffin and olive oil.

Notes
Hygroscopic. Incompatible with strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 3811-73-2
Molecular Formula C5H4NNaOS
Molecular Weight (g/mol) 149.143
MDL Number MFCD01941547
InChI Key XNRNJIIJLOFJEK-UHFFFAOYSA-N
Synonym sodium omadine, sodium pyrithione, sodium 2-pyridylthio-n-oxide, pyrithione sodium salt, thione reagent, 2-pyridinethiol-1-oxide sodium salt, 2-mercaptopyridine n-oxide sodium salt, omadine sodium, sodium-2-pyridinethiol-1-oxide, sodium omadine van
PubChem CID 19658
IUPAC Name sodium;1-oxidopyridine-2-thione
SMILES C1=CC(=S)N(C=C1)[O-].[Na+]

Specifications

Melting Point ∼290°C (decomposition)
Quantity 1 g
Sensitivity Hygroscopic
Merck Index 14,7994
Solubility Information Soluble in water,ethanol,propylene glycol,polyethylene glycol and dimethyl sulfoxide. Insoluble in liquid paraffin and olive oil.
Formula Weight 149.15
Percent Purity 98%
Chemical Name or Material 2-Mercaptopyridine N-oxide sodium salt, anhydrous

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.