missing translation for 'onlineSavingsMsg'
Learn More

2-Mercaptopyridine N-oxide sodium salt, 40% aq. soln.

Product Code. 11418756
Click to view available options
Quantity:
100 mL
500 mL
2500 mL
Unit Size:
100mL
2500mL
500mL
This item is not returnable. View return policy

Product Code. 11418756

Brand: Thermo Scientific Alfa Aesar A16922.0F

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 3811-73-2 | C5H4NNaOS | 149.143 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Suitable for battery materials development.

Applications
2-Mercaptopyridine N-oxide sodium salt is one of the active components in paint, sealants, shampoo, adhesive and aerosol due to its anti-microbial activity. In biochemistry studies, it is utilized to transport zinc into cells. Further, it is used to form bidentate oxothiolane chelates with transition metals. It acts as a stabilizer and viscosity building provider in weak basic or neutral medium.

Solubility
Miscible with ethanol, propylene glycol, polyethylene glycol and dimethyl sulfoxide. Immiscible with liquid paraffin and olive oil.

Notes
Hygroscopic. Incompatible with strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 3811-73-2
Molecular Formula C5H4NNaOS
Molecular Weight (g/mol) 149.143
MDL Number MFCD01941547
InChI Key XNRNJIIJLOFJEK-UHFFFAOYSA-N
Synonym sodium omadine, sodium pyrithione, sodium 2-pyridylthio-n-oxide, pyrithione sodium salt, thione reagent, 2-pyridinethiol-1-oxide sodium salt, 2-mercaptopyridine n-oxide sodium salt, omadine sodium, sodium-2-pyridinethiol-1-oxide, sodium omadine van
PubChem CID 19658
IUPAC Name sodium;1-oxidopyridine-2-thione
SMILES C1=CC(=S)N(C=C1)[O-].[Na+]

Specifications

Density 1.238
Refractive Index 1.4825
Quantity 2500 mL
Beilstein 4026050
Merck Index 14,7994
Solubility Information Miscible with ethanol,propylene glycol,polyethylene glycol and dimethyl sulfoxide. Immiscible with liquid paraffin and olive oil.
Formula Weight 149.15
Concentration or Composition (by Analyte or Components) 40% aq. soln.
Chemical Name or Material 2-Mercaptopyridine N-oxide sodium salt

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.