missing translation for 'onlineSavingsMsg'
Learn More

2-Mercaptopyridine N-oxide, 99%

Product Code. 11428136
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
Unit Size:
100g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11428136 100 g 100g
11408136 5 g 5g
11418136 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11428136 Supplier Thermo Scientific Alfa Aesar Supplier No. A14152.22
 Restricted Item
Estimated Shipment: 11-08-2026
Add to basket
This item is not returnable. View return policy

CAS: 1121-31-9 | C5H5NOS | 127.161 g/mol

2-Mercaptopyridine N-oxide is used in ratiometric fluorescence imaging of intracellular zinc ions in mammalian cultures. It is used in the radical coupling of benzoyl derivatives of 2-deoxy-D-ribose, with heterocyclic bases to give C-nucleosides. It acts as a ligand and form coordination complexes with palladium chloride. It is a precursor to O-acyl thiohydroxamates. Further, it serves as an antifungal agent. In addition to this, it is used in the preparation of 1-oxa-2-oxo-3-thia-indolizinium chloride by reaction with carbonyl dichloride.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-Mercaptopyridine N-oxide is used in ratiometric fluorescence imaging of intracellular zinc ions in mammalian cultures. It is used in the radical coupling of benzoyl derivatives of 2-deoxy-D-ribose, with heterocyclic bases to give C-nucleosides. It acts as a ligand and form coordination complexes with palladium chloride. It is a precursor to O-acyl thiohydroxamates. Further, it serves as an antifungal agent. In addition to this, it is used in the preparation of 1-oxa-2-oxo-3-thia-indolizinium chloride by reaction with carbonyl dichloride.

Solubility
Soluble in tris-hydrochloric acid.

Notes
Incompatible with strong oxidizing agents and strong bases.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1121-31-9
Molecular Formula C5H5NOS
Molecular Weight (g/mol) 127.161
MDL Number MFCD00006196
InChI Key YBBJKCMMCRQZMA-UHFFFAOYSA-N
Synonym pyrithione, 2-mercaptopyridine n-oxide, 2-mercaptopyridine-n-oxide, 1-hydroxy-2-pyridinethione, omadine, 2-pyridinethiol, 1-oxide, 1-hydroxyl-1h-pyridine-2-thione, n-hydroxypyridine-2-thione, 1-hydroxy-2 1h-pyridinethione, 2-pyridinethiol 1-oxide
PubChem CID 1570
ChEBI CHEBI:36578
IUPAC Name 1-hydroxypyridine-2-thione
SMILES C1=CC(=S)N(C=C1)O

Specifications

Melting Point 69°C to 72°C
Quantity 100 g
Beilstein 906983
Sensitivity Air and light sensitive
Merck Index 14,7994
Solubility Information Soluble in tris-hydrochloric acid.
Formula Weight 127.17
Percent Purity 99%
Chemical Name or Material 2-Mercaptopyridine N-oxide

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.