missing translation for 'onlineSavingsMsg'
Learn More

2-Mercaptopropionic acid, 97%

Product Code. 11378325
Change view
Click to view available options
Quantity:
10 g
100 g
500 g
Unit Size:
100g
10g
500g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11378325 10 g 10g
11388325 100 g 100g
11398325 500 g 500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11378325 Supplier Thermo Scientific Alfa Aesar Supplier No. L10257.09
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 79-42-5 | C3H6O2S | 106.14 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 79-42-5
Molecular Formula C3H6O2S
Molecular Weight (g/mol) 106.14
MDL Number MFCD00004862
InChI Key PMNLUUOXGOOLSP-UHFFFAOYNA-N
Synonym 2-mercaptopropionic acid, thiolactic acid, 2-mercaptopropanoic acid, 2-thiolactic acid, 2-thiolpropionic acid, alpha-mercaptopropionic acid, alpha-mercaptopropanoic acid, propionic acid, 2-mercapto, propanoic acid, 2-mercapto, fema no. 3180
PubChem CID 62326
ChEBI CHEBI:47872
IUPAC Name 2-sulfanylpropanoic acid
SMILES CC(S)C(O)=O

Specifications

Melting Point 10°C to 14°C
Density 1.196
Boiling Point 217°C
Flash Point 107°C (225°F)
Odor Unpleasant
Refractive Index 1.482
Quantity 10 g
UN Number UN2936
Beilstein 506218
Merck Index 14,9340
Formula Weight 106.14
Percent Purity 97%
Chemical Name or Material 2-Mercaptopropionic acid
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.