missing translation for 'onlineSavingsMsg'
Learn More

2-Mercaptoisobutyric Acid, 90%, TRC

Product Code. 30010857
Change view
Click to view available options
Quantity:
100 mg
1 g
10 g
Unit Size:
100mg
10g
1g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30010857 100 mg 100mg
30132733 10 g 10g
30134269 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30010857 Supplier Toronto Research Chemicals Supplier No. TRCM253400100MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 4695-31-2
InChI Formula InChI=1S/C4H8O2S/c1-4(2,7)3(5)6/h7H,1-2H3,(H,5,6)
IUPAC Name 2-methyl-2-sulfanylpropanoic acid
Molecular Formula C4 H8 O2 S
Molecular Weight (g/mol) 120.17
Chemical Name or Material 2-Mercaptoisobutyric Acid

Specifications

Quantity 100 mg
SMILES CC(C)(S)C(=O)O
Formula Weight 120.0245
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.