missing translation for 'onlineSavingsMsg'
Learn More

2-Mercaptoethanol, TRC

Product Code. 30467616
Change view
Click to view available options
Quantity:
500 g
Unit Size:
500g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30467616 500 g 500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30467616 Supplier Toronto Research Chemicals Supplier No. TRCM225480500G
 Restricted Item

This item requires a price update prior to being ordered.

Request a Price for this item here.

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 2-Mercaptoethanol
CAS 60-24-2
Molecular Formula C2 H6 O S
Quantity 500 g
Synonym 2-Mercaptoethanol, 1-Hydroxy-2-mercaptoethane, 1-Mercapto-2-hydroxyethane, 2-Hydroxy-1-ethanethiol, 2-Hydroxyethanethiol, 2-Hydroxyethyl mercaptan, 2-ME, 2-Mercapto-1-ethanol, 2-Mercaptoethyl alcohol, 2-Sulfanylethan-1-ol, 2-Sulfanylethanol, 2BME, BME, Beta-mercapto, Beta-mercaptoethanol, Ethylene glycol, monothio-, Hydroxyethyl mercaptan, Mercaptoethanol, Monothioethylene glycol, Monothioglycol, NSC 3723, Thioethylene glycol, Thiomonoglycol, β-Hydroxyethanethiol, β-Hydroxyethylmercaptan, β-Mercapto, β-Mercaptoethanol, β-met
InChI Formula InChI=1S/C2H6OS/c3-1-2-4/h3-4H,1-2H2
SMILES OCCS
IUPAC Name 2-sulfanylethanol
Molecular Weight (g/mol) 78.133
Formula Weight 78.0139
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.