missing translation for 'onlineSavingsMsg'
Learn More

2-Mercaptoethanol (Electrophoresis), Fisher BioReagents

Product Code. 10366313
Change view
Click to view available options
Quantity:
89.74 mL
Packaging:
Amber Glass/Poison Pack
Unit Size:
100g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10366313 89.74 mL Amber Glass/Poison Pack 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10366313 Supplier Fisher BioReagents Supplier No. BP176100
  • ONLINE EXCLUSIVE PRICE
    £68.55
    £66.00 / 100g
    Save £2.55
    3% Off
Request Bulk or Custom Formats

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

Currently Unavailable: We have no information when this product will be available.
This item is not returnable. View return policy
2-Mercaptoethanol reduces disulfide linkages in proteins and peptides and is used in many enzymatic reactions as a protein stabilizer.
TRUSTED_SUSTAINABILITY

Specifications

Alpha Vector Mercaptoethanol
CAS 60-24-2
Absorbance 0.05 AU max.
Melting Point -100°C
Color Yellow
Density 1.110g/cm³
pH 5.2
Boiling Point 157°C
Assay Percent Range ≥98 %
Packaging Amber Glass/Poison Pack
Molecular Formula C2H6OS
Linear Formula HSCH2CH2OH
MDL Number MFCD00004890
Quantity 89.74 mL
Merck Index 15, 5936
Synonym 2-mercaptoethanol, mercaptoethanol, thioglycol, beta-mercaptoethanol, ethanol, 2-mercapto, 2-thioethanol, 2-hydroxy-1-ethanethiol, thioethylene glycol, 2-hydroxyethanethiol, 2-hydroxyethyl mercaptan
InChI Key DGVVWUTYPXICAM-UHFFFAOYSA-N
SMILES OCCS
IUPAC Name 2-sulfanylethanol
Molecular Weight (g/mol) 78.13
PubChem CID 1567
Vapor Density 2.69g/mol
ChEBI CHEBI:41218
Formula Weight 78.13
Percent Purity ≥98%
Grade Electrophoresis
Physical Form Liquid
Chemical Name or Material 2-Mercaptoethanol
Show More Show Less
compliance-icons compliance-icons compliance-icons compliance-icons
Product Identifier
  • 2-Mercaptoethanol
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 3
  • Acute toxicity Category 2
  • Skin corrosion/irritation Category 2
  • Serious eye damage/eye irritation Category 1
  • Skin sensitiser Category 1A
  • Specific target organ toxicity after repeated exposure Category 2
  • LONG-TERM AQUATIC HAZARD Chronic 1
Hazard Statement
  • H301-Toxic if swallowed.
  • H310-Fatal in contact with skin.
  • H331-Toxic if inhaled.
  • H315-Causes skin irritation.
  • H318-Causes serious eye damage.
  • H317-May cause an allergic skin reaction.
  • H373-May cause damage to organs.
  • H410-Very toxic to aquatic life with long lasting effects.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.