missing translation for 'onlineSavingsMsg'
Learn More

2-Mercaptoethanol, 99%, pure

Product Code. 10265220 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
10 mL
250 mL
1 L
2.5 L
Packaging:
Glass bottle
Unit Size:
1 L
10mL
2.5 L
250mL
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10265220 10 mL Glass bottle 10mL
10367100 250 mL Glass bottle 250mL
10407520 1 L Glass bottle 1 L
11985721 2.5 L Glass bottle 2.5 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 10265220 Supplier Thermo Scientific Acros Supplier No. 125470100
  • ONLINE EXCLUSIVE PRICE
    £31.00
    £27.65 / 10mL
    Save £3.35
    10% Off
 Restricted Item
In Stock
Add to basket
This item is not returnable. View return policy

CAS: 60-24-2 | C2H6OS | 78.13 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 60-24-2
Molecular Formula C2H6OS
Molecular Weight (g/mol) 78.13
MDL Number MFCD00004890
InChI Key DGVVWUTYPXICAM-UHFFFAOYSA-N
Synonym 2-mercaptoethanol, mercaptoethanol, thioglycol, beta-mercaptoethanol, ethanol, 2-mercapto, 2-thioethanol, 2-hydroxy-1-ethanethiol, thioethylene glycol, 2-hydroxyethanethiol, 2-hydroxyethyl mercaptan
PubChem CID 1567
ChEBI CHEBI:41218
SMILES OCCS

Specifications

Melting Point -100.0°C
Color Colorless to Yellow
Density 1.1100g/mL
Boiling Point 157.0°C to 158.0°C
Flash Point 73°C
Infrared Spectrum Authentic
Assay Percent Range 98.5% min. (GC)
Packaging Glass bottle
Linear Formula HSCH2CH2OH
Refractive Index 1.4990 to 1.5020
Quantity 10 mL
Beilstein 01,47
Fieser 01,643
Merck Index 15,5936
Specific Gravity 1.11
Solubility Information Solubility in water: soluble. Other solubilities: miscible with alcohol,ether,benzene,soluble in acetone,miscible with most common organic solvents
IUPAC Name 2-sulfanylethanol
Viscosity 3.43 mPa.s (20°C)
Formula Weight 78.13
Percent Purity 99%
Grade Pure
Physical Form Liquid
Chemical Name or Material 2-Mercaptoethanol
Show More Show Less
compliance-icons compliance-icons compliance-icons compliance-icons
Product Identifier
  • 2-Mercaptoethanol
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 3
  • Acute toxicity Category 2
  • Skin corrosion/irritation Category 2
  • Serious eye damage/eye irritation Category 1
  • Skin sensitiser Category 1A
  • Specific target organ toxicity after repeated exposure Category 2
  • LONG-TERM AQUATIC HAZARD Chronic 1
Hazard Statement
  • H301-Toxic if swallowed.
  • H310-Fatal in contact with skin.
  • H331-Toxic if inhaled.
  • H315-Causes skin irritation.
  • H318-Causes serious eye damage.
  • H317-May cause an allergic skin reaction.
  • H373-May cause damage to organs.
  • H410-Very toxic to aquatic life with long lasting effects.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.