missing translation for 'onlineSavingsMsg'
Learn More

2-Mercaptoethane sulfonic acid sodium salt, 96%

Product Code. 15496129
Change view
Click to view available options
Quantity:
10 g
25 g
Unit Size:
10g
25g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15496129 25 g 25g
15486129 10 g 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15496129 Supplier Thermo Scientific Alfa Aesar Supplier No. J63989.14
 Restricted Item
Estimated Shipment: 07-12-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 19767-45-4 | C2H5NaO3S2 | 164.169 g/mol

2-Mercaptoethane sulfonic acid sodium salt is used as an adjuvant in cancer chemotherapy involving cyclophosphamide and ifosfamide. It is used as an antioxidant and cytoprotective agent in a wide variety of applications from protection from toxicity of therapeutic agents, such as cyclophosphamide to prevention from brain injury damage.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-Mercaptoethane sulfonic acid sodium salt is used as an adjuvant in cancer chemotherapy involving cyclophosphamide and ifosfamide. It is used as an antioxidant and cytoprotective agent in a wide variety of applications from protection from toxicity of therapeutic agents, such as cyclophosphamide to prevention from brain injury damage.

Solubility
Soluble in water. Also soluble in methanol, DMSO (33 mg/ml) and ethanol (<1mg/mL).

Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 19767-45-4
Molecular Formula C2H5NaO3S2
Molecular Weight (g/mol) 164.169
MDL Number MFCD00007535
InChI Key XOGTZOOQQBDUSI-UHFFFAOYSA-M
Synonym mesna, sodium 2-mercaptoethanesulfonate, mesnex, uromitexan, mitexan, 2-mercaptoethanesulfonic acid sodium salt, mistabron, mesnum, mistabronco, mucofluid
PubChem CID 23662354
IUPAC Name sodium;2-sulfanylethanesulfonate
SMILES C(CS(=O)(=O)[O-])S.[Na+]

Specifications

Color White
Linear Formula HSCH2CH2SO3Na
Quantity 25 g
Beilstein 3657828
Merck Index 14,5909
Solubility Information Soluble in water. Also soluble in methanol,DMSO (33mg/ml) and ethanol (<1mg/ml).
Formula Weight 164.18
Percent Purity 96%
Physical Form Powder
Chemical Name or Material 2-Mercaptoethane sulfonic acid sodium salt

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.