missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2-Mercaptobenzothiazole, ≥99.0%, Honeywell Fluka™
for the determination of Ag, Au, Bi, Cd, Hg, Ir, Pt, Tl,≥99.0%
£62.54 - £336.95
Chemical Identifiers
CAS | 149-30-4 |
---|---|
Molecular Formula | C7H5NS2 |
Molecular Weight (g/mol) | 167.244 |
MDL Number | MFCD00005781 |
InChI Key | YXIWHUQXZSMYRE-UHFFFAOYSA-N |
Synonym | 2-mercaptobenzothiazole, 2-benzothiazolethiol, captax, benzothiazolethiol, 2 3h-benzothiazolethione, benzothiazole-2-thiol, 1,3-benzothiazole-2-thiol, benzo d thiazole-2-thiol, rokon, rotax |
PubChem CID | 697993 |
ChEBI | CHEBI:34292 |
IUPAC Name | 3H-1,3-benzothiazole-2-thione |
SMILES | C1=CC=C2C(=C1)NC(=S)S2 |
Description
- Leading the Industry For Over 65 Years
- Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
Chemical Identifiers
149-30-4 | |
167.244 | |
YXIWHUQXZSMYRE-UHFFFAOYSA-N | |
697993 | |
3H-1,3-benzothiazole-2-thione |
C7H5NS2 | |
MFCD00005781 | |
2-mercaptobenzothiazole, 2-benzothiazolethiol, captax, benzothiazolethiol, 2 3h-benzothiazolethione, benzothiazole-2-thiol, 1,3-benzothiazole-2-thiol, benzo d thiazole-2-thiol, rokon, rotax | |
CHEBI:34292 | |
C1=CC=C2C(=C1)NC(=S)S2 |
Specifications
149-30-4 | |
Yellow | |
No data | |
MFCD00005781 | |
119484 | |
YXIWHUQXZSMYRE-UHFFFAOYSA-N | |
3H-1,3-benzothiazole-2-thione | |
697993 | |
167.25g/mol | |
Powder |
179°C to 182°C | |
260°C | |
C7H5NS2 | |
UN3077 | |
2-mercaptobenzothiazole, 2-benzothiazolethiol, captax, benzothiazolethiol, 2 3h-benzothiazolethione, benzothiazole-2-thiol, 1,3-benzothiazole-2-thiol, benzo d thiazole-2-thiol, rokon, rotax | |
C1=CC=C2C(=C1)NC(=S)S2 | |
167.244 | |
CHEBI:34292 | |
≥99.0% | |
2-Mercaptobenzothiazole |
Safety and Handling
P273-P280-P501
missing translation for 'dotInformation' : 9 / PGIII
missing translation for 'rtecsNumber' : 205-736-8